Assay Details
| Gene Symbol: USF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082635.1 | NP_001076104.1 | 5 - 6 | 403 | 62 | |
| XM_008263975.2 | XP_008262197.1 | 5 - 6 | 484 | 62 | ||
| XM_008263976.2 | XP_008262198.1 | 5 - 6 | 1048 | 62 | ||
| XM_008263977.2 | XP_008262199.1 | 5 - 6 | 483 | 62 | ||
| XM_008263978.2 | XP_008262200.1 | 3 - 4 | 238 | 62 | ||
| GenBank mRNA | AF003894.1 | - | 5 - 6 | 403 | 62 | |
| AF003895.1 | - | 5 - 6 | 403 | 62 | ||
More Information
Set Membership: |
|

USF1
Genomic Map