Assay Details
| Gene Symbol: ELP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082655.1 | NP_001076124.1 | 33 - 34 | 3705 | 87 | |
| GenBank mRNA | AF388202.1 | - | 33 - 34 | 3705 | 87 | |
More Information
Set Membership: |
|

ELP1
Genomic Map