Assay Details
| Gene Symbol: CD38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082683.1 | NP_001076152.1 | 3 - 4 | 571 | 66 | |
| XM_008274042.2 | XP_008272264.1 | 4 - 5 | 988 | 66 | ||
| XM_017350127.1 | XP_017205616.1 | 4 - 5 | 1020 | 66 | ||
| GenBank mRNA | AF272974.1 | - | 3 - 4 | 571 | 66 | |
More Information
Set Membership: |
|

CD38
Genomic Map