Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082685.2 | NP_001076154.1 | 3 - 4 | 614 | 79 | |
| GenBank mRNA | D49844.1 | - | 3 - 4 | 614 | 79 | |
More Information
Set Membership: |
|

CTLA4
Genomic Map