Assay Details
Gene Symbol: LGALS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082713.1 | NP_001076182.1 | 501 | 68 | ||
GenBank mRNA | AF091738.1 | - | 501 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
