Assay Details
| Gene Symbol: CBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082749.1 | NP_001076218.1 | 516 | 107 | ||
| GenBank mRNA | U07051.1 | - | 516 | 107 | ||
| U09244.1 | - | 480 | 107 | |||
More Information
Set Membership: |
|

CBR1
Genomic Map