Assay Details
| Gene Symbol: HPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082760.1 | NP_001076229.1 | 9 - 10 | 1119 | 85 | |
| GenBank mRNA | X16429.1 | - | 1119 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082760.1 | NP_001076229.1 | 9 - 10 | 1119 | 85 | |
| GenBank mRNA | X16429.1 | - | 1119 | 85 | ||
Set Membership: |
|