Assay Details
| Gene Symbol: SCNN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082775.1 | NP_001076244.1 | 1 - 2 | 350 | 61 | |
| GenBank mRNA | AJ132109.1 | - | 1 - 2 | 350 | 61 | |
More Information
Set Membership: |
|

SCNN1B
Genomic Map