Assay Details
| Gene Symbol: CD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089312.1 | NP_001082781.1 | 4 - 5 | 882 | 91 | |
| XM_008264000.2 | XP_008262222.1 | 3 - 4 | 1787 | 91 | ||
| XM_008264002.2 | XP_008262224.1 | 4 - 5 | 1316 | 91 | ||
| GenBank mRNA | AF276979.1 | - | 4 - 5 | 882 | 91 | |
More Information
Set Membership: |
|

CD1B
Genomic Map