Assay Details
| Gene Symbol: CLDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089316.1 | NP_001082785.1 | 1 - 2 | 227 | 93 | |
| GenBank mRNA | DQ993356.1 | - | 1 - 2 | 227 | 93 | |
More Information
Set Membership: |
|

CLDN1
Genomic Map