Assay Details
| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099966.1 | NP_001093436.1 | 5 - 6 | 757 | 99 | |
| XM_008258926.2 | XP_008257148.1 | 6 - 7 | 948 | 99 | ||
| XM_017342622.1 | XP_017198111.1 | 6 - 7 | 1016 | 99 | ||
| GenBank mRNA | EF397306.1 | - | 5 - 6 | 757 | 99 | |
More Information
Set Membership: |
|

CASP10
Genomic Map