Assay Details
| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101686.1 | NP_001095156.1 | 1469 | 69 | ||
| GenBank mRNA | X06087.1 | - | 1469 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP2CA| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101686.1 | NP_001095156.1 | 1469 | 69 | ||
| GenBank mRNA | X06087.1 | - | 1469 | 69 | ||
Set Membership: |
|