Assay Details
| Gene Symbol: APOA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101687.1 | NP_001095157.1 | 2 - 3 | 95 | 105 | |
| GenBank mRNA | DQ310702.1 | - | 1 - 2 | 43 | 105 | |
| X06658.1 | - | 95 | 105 | |||
| X15908.1 | - | 66 | 105 | |||
More Information
Set Membership: |
|

APOA1
Genomic Map