Assay Details
| Gene Symbol: TNFSF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105676.1 | NP_001099146.1 | 2 - 3 | 366 | 111 | |
| GenBank mRNA | EF494239.1 | - | 2 - 3 | 366 | 111 | |
More Information
Set Membership: |
|

TNFSF13
Genomic Map