Assay Details
| Gene Symbol: SLC2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105687.1 | NP_001099157.1 | 2 - 3 | 400 | 61 | |
| GenBank mRNA | M21747.1 | - | 2 - 3 | 400 | 61 | |
More Information
Set Membership: |
|

SLC2A1
Genomic Map