Assay Details
Gene Symbol: LOC100125995 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109819.1 | NP_001103289.1 | 870 | 56 | ||
GenBank mRNA | EF120627.1 | - | 870 | 56 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
