Assay Details
Gene Symbol: LOC100125995 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109819.1 | NP_001103289.1 | 941 | 60 | ||
XM_008250334.2 | XP_008248556.1 | 946 | 60 | |||
XM_008250335.2 | XP_008248557.1 | 946 | 60 | |||
XM_017338587.1 | XP_017194076.1 | 946 | 60 | |||
XM_017338589.1 | XP_017194078.1 | 946 | 60 | |||
GenBank mRNA | EF120627.1 | - | 941 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
