Assay Details
Gene Symbol: LOC100009266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128073.1 | NP_001121545.1 | 2 - 3 | 452 | 76 | |
GenBank mRNA | X14522.1 | - | 452 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
