Assay Details
Gene Symbol: LOC100008810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082104.2 | NP_001075573.1 | 349 | 92 | ||
XM_017350720.1 | XP_017206209.1 | 849 | 92 | |||
GenBank mRNA | AF346415.1 | - | 351 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
