Assay Details
| Gene Symbol: SLC9A3R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082107.1 | NP_001075576.1 | 1447 | 112 | ||
| GenBank mRNA | AF358433.1 | - | 1447 | 112 | ||
More Information
Set Membership: |
|

SLC9A3R2
Genomic Map