Assay Details
| Gene Symbol: CLIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082111.1 | NP_001075580.1 | 4 | 352 | 103 | |
| XM_008262536.2 | XP_008260758.2 | 4 | 334 | 103 | ||
| GenBank mRNA | AF387765.1 | - | 4 | 352 | 103 | |
More Information
Set Membership: |
|

CLIC1
Genomic Map