Assay Details
| Gene Symbol: TPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082129.2 | NP_001075598.1 | 6 | 1047 | 99 | |
| GenBank mRNA | AJ277093.1 | - | 6 | 1051 | 99 | |
| Gene Symbol: LOC100353956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ131951.1 | - | 6 | 1046 | 99 | |
| Gene Symbol: LOC100357892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ277092.1 | - | 6 | 1048 | 99 | |
More Information
Set Membership: |
|

TPT1
Genomic Map