Assay Details
| Gene Symbol: IL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082201.1 | NP_001075670.1 | 3 | 646 | 86 | |
| GenBank mRNA | D21835.1 | - | 3 | 646 | 86 | |
| KT216047.1 | - | 3 | 560 | 86 | ||
| M26295.1 | - | 3 | 620 | 86 | ||
More Information
Set Membership: |
|

IL1B
Genomic Map