Assay Details
| Gene Symbol: GAPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082253.1 | NP_001075722.1 | 5 | 346 | 82 | |
| GenBank mRNA | L23961.1 | - | 5 | 346 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAPDH| Gene Symbol: GAPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082253.1 | NP_001075722.1 | 5 | 346 | 82 | |
| GenBank mRNA | L23961.1 | - | 5 | 346 | 82 | |
Set Membership: |
|