Assay Details
| Gene Symbol: GRO-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082386.1 | NP_001075855.1 | 754 | 79 | ||
| GenBank mRNA | U95808.1 | - | 754 | 79 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRO-A| Gene Symbol: GRO-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082386.1 | NP_001075855.1 | 754 | 79 | ||
| GenBank mRNA | U95808.1 | - | 754 | 79 | ||
Set Membership: | 
		   
					
						
							 |