Assay Details
| Gene Symbol: SLC6A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082397.1 | NP_001075866.1 | 1947 | 85 | ||
| GenBank mRNA | X67252.1 | - | 1947 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC6A8| Gene Symbol: SLC6A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082397.1 | NP_001075866.1 | 1947 | 85 | ||
| GenBank mRNA | X67252.1 | - | 1947 | 85 | ||
Set Membership: |
|