Assay Details
Gene Symbol: HINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082623.1 | NP_001076092.1 | 1 | 147 | 72 | |
GenBank mRNA | Y11175.1 | - | 147 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
