Assay Details
| Gene Symbol: KCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082717.1 | NP_001076186.1 | 7 | 1855 | 66 | |
| XM_017345496.1 | XP_017200985.1 | 6 | 1873 | 66 | ||
| GenBank mRNA | AF198445.1 | - | 7 | 1855 | 66 | |
| AF493549.1 | - | 6 | 1584 | 66 | ||
More Information
Set Membership: |
|

KCND3
Genomic Map