Assay Details
| Gene Symbol: ACTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101683.1 | NP_001095153.1 | 1 | 1651 | 106 | |
| GenBank mRNA | X60733.1 | - | 1651 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACTB| Gene Symbol: ACTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101683.1 | NP_001095153.1 | 1 | 1651 | 106 | |
| GenBank mRNA | X60733.1 | - | 1651 | 106 | ||
Set Membership: |
|