Assay Details
| Gene Symbol: CAB39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712517.3 | XP_002712563.1 | 3 - 4 | 503 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAB39| Gene Symbol: CAB39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712517.3 | XP_002712563.1 | 3 - 4 | 503 | 69 | |
Set Membership: |
|