Assay Details
| Gene Symbol: LDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195707.1 | NP_001182636.1 | 3 - 4 | 427 | 64 | |
| XM_008259339.2 | XP_008257561.1 | 4 - 5 | 573 | 64 | ||
| GenBank mRNA | M22584.1 | - | 1 - 2 | 70 | 64 | |
More Information
Set Membership: |
|

LDHB
Genomic Map