Assay Details
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082744.2 | NP_001076213.1 | 4 - 5 | 662 | 93 | |
GenBank mRNA | L20858.1 | - | 4 - 5 | 660 | 93 | |
M95581.1 | - | 4 - 5 | 397 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
