Assay Details
| Gene Symbol: AQP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344416.1 | XP_017199905.1 | 1 - 2 | 461 | 94 | |
| GenBank mRNA | AF495880.1 | - | 1 - 2 | 322 | 94 | |
More Information
Set Membership: |
|

AQP1
Genomic Map