Assay Details
| Gene Symbol: CKMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163070.1 | NP_001156542.1 | 1 - 2 | 182 | 72 | |
| GenBank mRNA | AJ011334.1 | - | 1 - 2 | 182 | 72 | |
More Information
Set Membership: |
|

CKMT2
Genomic Map