Assay Details
| Gene Symbol: TAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082165.1 | NP_001075634.1 | 1 - 2 | 130 | 88 | |
| GenBank mRNA | AY471576.1 | - | 1 - 2 | 130 | 88 | |
| AY471577.1 | - | 1 - 2 | 130 | 88 | ||
More Information
Set Membership: |
|

TAC4
Genomic Map