Assay Details
| Gene Symbol: AICDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343607.1 | XP_017199096.1 | 2 - 3 | 182 | 67 | |
| GenBank mRNA | AY928183.1 | - | 1 - 2 | 138 | 67 | |
More Information
Set Membership: |
|

AICDA
Genomic Map