Assay Details
| Gene Symbol: NPY4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160280.1 | NP_001153752.1 | 1 | 350 | 67 | |
| XM_008269879.2 | XP_008268101.1 | 2 | 506 | 67 | ||
| GenBank mRNA | AB469825.1 | - | 1 | 350 | 67 | |
More Information
Set Membership: |
|

NPY4R
Genomic Map