Assay Details
| Gene Symbol: SNURF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082714.1 | NP_001076183.1 | 1 - 2 | 281 | 83 | |
| GenBank mRNA | AF101043.1 | - | 1 - 2 | 281 | 83 | |
More Information
Set Membership: |
|

SNURF
Genomic Map