Assay Details
| Gene Symbol: HSD17B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719167.3 | XP_002719213.1 | 2 - 3 | 837 | 71 | |
| GenBank mRNA | AF265661.1 | - | 1 - 2 | 86 | 71 | |
More Information
Set Membership: |
|

HSD17B1
Genomic Map