Assay Details
| Gene Symbol: CDKN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712662.3 | XP_002712708.1 | 2 - 3 | 1240 | 86 | |
| XM_017343370.1 | XP_017198859.1 | 2 - 3 | 1322 | 86 | ||
| GenBank mRNA | DQ845181.1 | - | 2 - 3 | 322 | 85 | |
More Information
Set Membership: |
|

CDKN1B
Genomic Map