Assay Details
Gene Symbol: LOC100144340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716358.3 | XP_002716404.1 | 1 | 574 | 77 | |
GenBank mRNA | EF222473.1 | - | 3 - 4 | 386 | 77 |
Gene Symbol: LOC100346276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712764.3 | XP_002712810.1 | 1 | 564 | 77 |
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713098.3 | XP_002713144.1 | 4 - 5 | 664 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
