Assay Details
| Gene Symbol: RARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719354.3 | XP_002719400.1 | 2 - 3 | 88 | ||
| GenBank mRNA | DQ102471.1 | - | 1 - 2 | 71 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RARA| Gene Symbol: RARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719354.3 | XP_002719400.1 | 2 - 3 | 88 | ||
| GenBank mRNA | DQ102471.1 | - | 1 - 2 | 71 | 88 | |
Set Membership: |
|