Assay Details
Gene Symbol: NPY5R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160285.2 | NP_001153757.1 | 1245 | 77 | ||
XM_008273474.2 | XP_008271696.1 | 1461 | 77 | |||
XM_008273476.2 | XP_008271698.1 | 1451 | 77 | |||
XM_008273477.2 | XP_008271699.1 | 1537 | 77 | |||
GenBank mRNA | AB469826.1 | - | 1250 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
