Assay Details
Gene Symbol: ADH2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170871.1 | NP_001164342.1 | 7 - 8 | 1123 | 101 | |
GenBank mRNA | AJ002388.1 | - | 7 - 8 | 1123 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
