Assay Details
| Gene Symbol: COX6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170883.2 | NP_001164354.1 | 2 - 3 | 289 | 98 | |
| GenBank mRNA | AF198089.1 | - | 2 - 3 | 293 | 98 | |
More Information
Set Membership: |
|

COX6A1
Genomic Map