Assay Details
Gene Symbol: PIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171116.1 | NP_001164587.1 | 2 - 3 | 204 | 96 | |
XM_017342637.1 | XP_017198126.1 | 2 - 3 | 257 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
