Assay Details
Gene Symbol: CCKBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171123.2 | NP_001164594.2 | 1 - 2 | 547 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
