Assay Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171278.1 | NP_001164749.1 | 2 - 3 | 154 | 141 | |
XM_008258181.2 | XP_008256403.1 | 2 - 3 | 367 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
