Assay Details
| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171278.1 | NP_001164749.1 | 3 - 4 | 300 | 84 | |
| XM_008258181.2 | XP_008256403.1 | 3 - 4 | 513 | 84 | ||
More Information
Set Membership: |
|

ZNF800
Genomic Map