Assay Details
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171288.1 | NP_001164759.1 | 1 - 2 | 166 | 89 | |
XM_008261390.2 | XP_008259612.1 | 3 - 4 | 396 | 89 | ||
XM_008261391.2 | XP_008259613.1 | 3 - 4 | 371 | 89 | ||
XM_008261392.2 | XP_008259614.1 | 3 - 4 | 367 | 89 | ||
XM_008261393.2 | XP_008259615.1 | 3 - 4 | 343 | 89 | ||
XM_008261394.2 | XP_008259616.1 | 2 - 3 | 851 | 89 | ||
XM_017343656.1 | XP_017199145.1 | 3 - 4 | 839 | 89 | ||
XM_017343657.1 | XP_017199146.1 | 3 - 4 | 814 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
